Assay Details
| Gene Symbol: atp5f1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024429.2 | NP_001019600.2 | 3 - 4 | 534 | 92 | |
| GenBank mRNA | AW281383.1 | - | 3 - 4 | 443 | 92 | |
| BC095620.1 | - | 3 - 4 | 471 | 92 | ||
| BC154659.1 | - | 3 - 4 | 455 | 92 | ||
| EH484343.1 | - | 3 - 4 | 500 | 92 | ||
More Information
Set Membership: |
|

atp5f1b
Genomic Map