Assay Details
| Gene Symbol: spra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024430.2 | NP_001019601.2 | 1 - 2 | 367 | 58 | |
| GenBank mRNA | AW420062.1 | - | 1 - 2 | 303 | 58 | |
| BC076547.1 | - | 1 - 2 | 305 | 58 | ||
More Information
Set Membership: |
|

spra
Genomic Map