Assay Details
| Gene Symbol: spra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024430.2 | NP_001019601.2 | 2 - 3 | 653 | 57 | |
| GenBank mRNA | BC076547.1 | - | 2 - 3 | 591 | 57 | |
More Information
Set Membership: |
|

spra
Genomic Map