Assay Details
| Gene Symbol: eif1axb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025453.1 | NP_001020624.1 | 2 - 3 | 233 | 74 | |
| GenBank mRNA | BC096794.1 | - | 2 - 3 | 233 | 74 | |
| BC155227.1 | - | 2 - 3 | 230 | 74 | ||
More Information
Set Membership: |
|

eif1axb
Genomic Map