Assay Details
| Gene Symbol: fdps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025471.1 | NP_001020642.1 | 5 - 6 | 570 | 86 | |
| GenBank mRNA | BC083515.1 | - | 4 - 5 | 501 | 86 | |
| BC097112.1 | - | 5 - 6 | 570 | 86 | ||
| BC164398.1 | - | 4 - 5 | 526 | 86 | ||
More Information
Set Membership: |
|

fdps
Genomic Map