Assay Details
| Gene Symbol: idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025475.1 | NP_001020646.1 | 2 - 3 | 471 | 143 | |
| XM_005162508.4 | XP_005162565.1 | 2 - 3 | 538 | 143 | ||
| GenBank mRNA | BC076541.1 | - | 1 - 2 | 77 | 143 | |
| BC097193.1 | - | 2 - 3 | 471 | 143 | ||
More Information
Set Membership: |
|

idi1
Genomic Map