Assay Details
| Gene Symbol: fbxo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025499.1 | NP_001020670.1 | 3 - 4 | 333 | 56 | |
| XM_021475037.1 | XP_021330712.1 | 3 - 4 | 364 | 56 | ||
| GenBank mRNA | BC097156.1 | - | 3 - 4 | 333 | 56 | |
More Information
Set Membership: |
|

fbxo7
Genomic Map