Assay Details
| Gene Symbol: hrh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025518.1 | NP_001020689.1 | 1 - 2 | 343 | 100 | |
| GenBank mRNA | BC162722.1 | - | 1 - 2 | 370 | 100 | |
| BC162728.1 | - | 1 - 2 | 370 | 100 | ||
| DQ647807.1 | - | 1 - 2 | 343 | 100 | ||
More Information
Set Membership: |
|

hrh3
Genomic Map