Assay Details
| Gene Symbol: podxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045016.2 | NP_001038481.1 | 7 - 8 | 1404 | 73 | |
| GenBank mRNA | BC133070.1 | - | 7 - 8 | 1404 | 73 | |
| BC163499.1 | - | 7 - 8 | 1319 | 73 | ||
| BC163515.1 | - | 7 - 8 | 1319 | 73 | ||
More Information
Set Membership: |
|

podxl
Genomic Map