Assay Details
| Gene Symbol: atf7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025549.1 | NP_001020720.1 | 6 - 7 | 684 | 91 | |
| XM_005166049.3 | XP_005166106.1 | 7 - 8 | 806 | 91 | ||
| XM_005166050.4 | XP_005166107.1 | 7 - 8 | 1169 | 91 | ||
| XM_005166051.3 | XP_005166108.1 | 7 - 8 | 806 | 91 | ||
| GenBank mRNA | AY928941.1 | - | 6 - 7 | 684 | 91 | |
More Information
Set Membership: |
|

atf7b
Genomic Map