Assay Details
| Gene Symbol: atf7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025549.1 | NP_001020720.1 | 7 - 8 | 808 | 69 | |
| XM_005166051.3 | XP_005166108.1 | 8 - 9 | 930 | 69 | ||
| GenBank mRNA | AY928941.1 | - | 7 - 8 | 808 | 69 | |
| BC163601.1 | - | 8 - 9 | 898 | 69 | ||
| BC163602.1 | - | 8 - 9 | 898 | 69 | ||
More Information
Set Membership: |
|

atf7b
Genomic Map