Assay Details
| Gene Symbol: aldoca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029952.1 | NP_001025123.1 | 7 - 8 | 884 | 116 | |
| XM_009291564.3 | XP_009289839.1 | 7 - 8 | 116 | |||
| GenBank mRNA | BC098624.1 | - | 7 - 8 | 884 | 116 | |
More Information
Set Membership: |
|

aldoca
Genomic Map