Assay Details
| Gene Symbol: aldoca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029952.1 | NP_001025123.1 | 8 - 9 | 1082 | 64 | |
| XM_009291564.3 | XP_009289839.1 | 8 - 9 | 1041 | 64 | ||
| GenBank mRNA | BC098624.1 | - | 8 - 9 | 1082 | 64 | |
More Information
Set Membership: |
|

aldoca
Genomic Map