Assay Details
Gene Symbol: ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029962.2 | NP_001025133.2 | 8 - 9 | 774 | 67 | |
XM_009307800.2 | XP_009306075.1 | 6 - 7 | 525 | 67 | ||
GenBank mRNA | BC097146.1 | - | 8 - 9 | 757 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
