Assay Details
| Gene Symbol: rgs7bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030069.1 | NP_001025240.1 | 2 - 3 | 517 | 81 | |
| GenBank mRNA | BC096790.1 | - | 2 - 3 | 517 | 81 | |
| BC153404.1 | - | 2 - 3 | 373 | 81 | ||
| BC165928.1 | - | 2 - 3 | 348 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC153404.1 transcripts.
Set Membership: |
|

rgs7bpb
Genomic Map