Assay Details
| Gene Symbol: brms1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030102.2 | NP_001025273.1 | 1 - 2 | 505 | 69 | |
| XM_005158646.4 | XP_005158703.1 | 1 - 2 | 483 | 69 | ||
| XM_005158647.4 | XP_005158704.1 | 1 - 2 | 483 | 69 | ||
| XM_009292865.3 | XP_009291140.1 | 1 - 2 | 484 | 69 | ||
| XM_009292866.3 | XP_009291141.1 | 1 - 2 | 484 | 69 | ||
| XM_021467237.1 | XP_021322912.1 | 1 - 2 | 484 | 69 | ||
| GenBank mRNA | BC097187.1 | - | 1 - 2 | 513 | 69 | |
| EH594471.1 | - | 1 - 2 | 491 | 69 | ||
More Information
Set Membership: |
|

brms1la
Genomic Map