Assay Details
| Gene Symbol: brms1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030102.2 | NP_001025273.1 | 9 - 10 | 1222 | 96 | |
| XM_005158646.4 | XP_005158703.1 | 9 - 10 | 1188 | 96 | ||
| XM_005158647.4 | XP_005158704.1 | 10 - 11 | 1214 | 96 | ||
| XM_021467237.1 | XP_021322912.1 | 10 - 11 | 1203 | 96 | ||
| GenBank mRNA | BC097187.1 | - | 9 - 10 | 1230 | 96 | |
| EB900863.1 | - | 4 - 5 | 244 | 96 | ||
More Information
Set Membership: |
|

brms1la
Genomic Map