Assay Details
| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 14 - 15 | 1681 | 92 | |
| XM_005170181.4 | XP_005170238.1 | 15 - 16 | 1782 | 92 | ||
| XM_021468448.1 | XP_021324123.1 | 16 - 17 | 1818 | 92 | ||
| XM_021468449.1 | XP_021324124.1 | 15 - 16 | 1737 | 92 | ||
| GenBank mRNA | BC115194.1 | - | 16 - 17 | 1847 | 92 | |
| BC163586.1 | - | 14 - 15 | 1500 | 92 | ||
| BC163590.1 | - | 14 - 15 | 1500 | 92 | ||
| BC167965.1 | - | 14 - 15 | 1662 | 92 | ||
More Information
Set Membership: |
|

snx9b
Genomic Map