Assay Details
| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 4 - 5 | 549 | 113 | |
| XM_021468449.1 | XP_021324124.1 | 4 - 5 | 569 | 113 | ||
| GenBank mRNA | BC163586.1 | - | 4 - 5 | 368 | 113 | |
| BC163590.1 | - | 4 - 5 | 368 | 113 | ||
| BC167965.1 | - | 4 - 5 | 530 | 113 | ||
More Information
Set Membership: |
|

snx9b
Genomic Map