Assay Details
| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 5 - 6 | 712 | 63 | |
| XM_005170181.4 | XP_005170238.1 | 6 - 7 | 813 | 63 | ||
| GenBank mRNA | BC115194.1 | - | 6 - 7 | 796 | 63 | |
| BC163586.1 | - | 5 - 6 | 531 | 63 | ||
| BC163590.1 | - | 5 - 6 | 531 | 63 | ||
| BC167965.1 | - | 5 - 6 | 693 | 63 | ||
More Information
Set Membership: |
|

snx9b
Genomic Map