Assay Details
| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 9 - 10 | 1446 | 119 | |
| XM_005171778.4 | XP_005171835.1 | 10 - 11 | 1618 | 119 | ||
| XM_005171779.4 | XP_005171836.1 | 10 - 11 | 1615 | 119 | ||
| XM_021475966.1 | XP_021331641.1 | 8 - 9 | 1411 | 119 | ||
| GenBank mRNA | BC171681.1 | - | 9 - 10 | 1477 | 119 | |
| DQ840138.1 | - | 4 - 5 | 471 | 119 | ||
More Information
Set Membership: |
|

ehmt1a
Genomic Map