Assay Details
| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 11 - 12 | 1640 | 80 | |
| XM_005171778.4 | XP_005171835.1 | 12 - 13 | 1812 | 80 | ||
| XM_005171779.4 | XP_005171836.1 | 12 - 13 | 1809 | 80 | ||
| XM_021475966.1 | XP_021331641.1 | 10 - 11 | 1605 | 80 | ||
| GenBank mRNA | BC171681.1 | - | 11 - 12 | 1671 | 80 | |
More Information
Set Membership: |
|

ehmt1a
Genomic Map