Assay Details
| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 5 - 6 | 862 | 76 | |
| XM_005171778.4 | XP_005171835.1 | 6 - 7 | 1034 | 76 | ||
| XM_005171779.4 | XP_005171836.1 | 6 - 7 | 1034 | 76 | ||
| XM_021475966.1 | XP_021331641.1 | 4 - 5 | 827 | 76 | ||
| GenBank mRNA | BC171681.1 | - | 5 - 6 | 893 | 76 | |
More Information
Set Membership: |
|

ehmt1a
Genomic Map