Assay Details
| Gene Symbol: myh9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098177.2 | NP_001091647.2 | 29 - 30 | 4118 | 99 | |
| XM_005165694.4 | XP_005165751.1 | 30 - 31 | 4291 | 99 | ||
| XM_005165695.4 | XP_005165752.1 | 29 - 30 | 4270 | 99 | ||
| GenBank mRNA | BC139512.1 | - | 3 - 4 | 294 | 99 | |
More Information
Set Membership: |
|

myh9a
Genomic Map