Assay Details
| Gene Symbol: slc17a7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098755.1 | NP_001092225.1 | 5 - 6 | 659 | 68 | |
| GenBank mRNA | BC142603.1 | - | 5 - 6 | 659 | 68 | |
More Information
Set Membership: |
|

slc17a7a
Genomic Map