Assay Details
| Gene Symbol: mul1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098760.1 | NP_001092230.1 | 2 - 3 | 163 | 93 | |
| XM_005166206.4 | XP_005166263.1 | 2 - 3 | 156 | 93 | ||
| GenBank mRNA | BC142887.1 | - | 2 - 3 | 163 | 93 | |
More Information
Set Membership: |
|

mul1b
Genomic Map