Assay Details
| Gene Symbol: agr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099232.2 | NP_001092702.2 | 5 - 6 | 388 | 84 | |
| GenBank mRNA | BC142869.1 | - | 5 - 6 | 356 | 84 | |
| EH452502.1 | - | 5 - 6 | 417 | 84 | ||
| EH464266.1 | - | 3 - 4 | 470 | 84 | ||
| JX566722.1 | - | 4 - 5 | 289 | 84 | ||
More Information
Set Membership: |
|

agr1
Genomic Map