Assay Details
Gene Symbol: otol1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099741.1 | NP_001093211.1 | 3 - 4 | 569 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
