Assay Details
| Gene Symbol: ahsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100029.1 | NP_001093499.1 | 2 - 3 | 525 | 68 | |
| GenBank mRNA | BC124812.1 | - | 2 - 3 | 356 | 68 | |
| BC134837.1 | - | 2 - 3 | 357 | 68 | ||
| BC150351.1 | - | 2 - 3 | 371 | 68 | ||
More Information
Set Membership: |
|

ahsg2
Genomic Map