Assay Details
| Gene Symbol: ahsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100029.1 | NP_001093499.1 | 3 - 4 | 603 | 69 | |
| GenBank mRNA | BC150351.1 | - | 3 - 4 | 449 | 69 | |
More Information
Set Membership: |
|

ahsg2
Genomic Map