Assay Details
| Gene Symbol: ahsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100029.1 | NP_001093499.1 | 4 - 5 | 758 | 131 | |
| GenBank mRNA | BC124812.1 | - | 4 - 5 | 589 | 131 | |
| BC134837.1 | - | 4 - 5 | 590 | 131 | ||
| BC150351.1 | - | 4 - 5 | 604 | 131 | ||
More Information
Set Membership: |
|

ahsg2
Genomic Map