Assay Details
Gene Symbol: emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102389.2 | NP_001095859.2 | 4 - 5 | 433 | 97 | |
NM_001163918.1 | NP_001157390.1 | 4 - 5 | 433 | 97 | ||
GenBank mRNA | BC150420.1 | - | 4 - 5 | 408 | 97 | |
EH461036.1 | - | 4 - 5 | 676 | 97 | ||
EH531631.1 | - | 4 - 5 | 705 | 97 | ||
EH586841.1 | - | 4 - 5 | 459 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
