Assay Details
| Gene Symbol: znrf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102637.1 | NP_001096107.1 | 4 - 5 | 663 | 64 | |
| XM_005169979.4 | XP_005170036.1 | 4 - 5 | 798 | 64 | ||
| XM_017352172.2 | XP_017207661.1 | 4 - 5 | 801 | 64 | ||
| GenBank mRNA | BC150372.1 | - | 4 - 5 | 663 | 64 | |
More Information
Set Membership: |
|

znrf2a
Genomic Map