Assay Details
| Gene Symbol: npffr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105109.1 | NP_001098579.1 | 2 - 3 | 433 | 68 | |
| XM_005165326.4 | XP_005165383.1 | 2 - 3 | 641 | 68 | ||
More Information
Set Membership: |
|

npffr2a
Genomic Map