Assay Details
| Gene Symbol: anxa2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105600.1 | NP_001099070.1 | 10 - 11 | 843 | 81 | |
| GenBank mRNA | BC110093.1 | - | 11 - 12 | 873 | 81 | |
| BC153582.1 | - | 10 - 11 | 843 | 81 | ||
More Information
Set Membership: |
|

anxa2b
Genomic Map