Assay Details
| Gene Symbol: rps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001819.2 | NP_001001819.1 | 2 - 3 | 109 | 100 | |
| GenBank mRNA | AY561511.1 | - | 2 - 3 | 113 | 100 | |
| BC081516.1 | - | 2 - 3 | 109 | 100 | ||
More Information
Set Membership: |
|

rps15
Genomic Map