Assay Details
| Gene Symbol: popdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001848.1 | NP_001001848.1 | 1 - 2 | 615 | 110 | |
| GenBank mRNA | AY293116.1 | - | 1 - 2 | 615 | 110 | |
| BC162409.1 | - | 1 - 2 | 568 | 110 | ||
| BC162676.1 | - | 1 - 2 | 568 | 110 | ||
More Information
Set Membership: |
|

popdc3
Genomic Map