Assay Details
| Gene Symbol: ino80c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002108.2 | NP_001002108.2 | 1 - 2 | 332 | 70 | |
| GenBank mRNA | BC071494.1 | - | 1 - 2 | 323 | 70 | |
| EH554982.1 | - | 1 - 2 | 370 | 70 | ||
| EH565174.1 | - | 4 - 5 | 574 | 70 | ||
More Information
Set Membership: |
|

ino80c
Genomic Map