Assay Details
| Gene Symbol: tmx2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002113.1 | NP_001002113.1 | 7 - 8 | 820 | 89 | |
| GenBank mRNA | BC071481.1 | - | 7 - 8 | 820 | 89 | |
More Information
Set Membership: |
|

tmx2b
Genomic Map