Assay Details
| Gene Symbol: rbks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002117.1 | NP_001002117.1 | 5 - 6 | 402 | 62 | |
| GenBank mRNA | BC071473.1 | - | 5 - 6 | 402 | 62 | |
More Information
Set Membership: |
|

rbks
Genomic Map