Assay Details
| Gene Symbol: trnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002159.1 | NP_001002159.1 | 6 - 7 | 884 | 73 | |
| XM_005166030.4 | XP_005166087.1 | 6 - 7 | 926 | 73 | ||
| GenBank mRNA | AY648718.1 | - | 6 - 7 | 924 | 73 | |
| BC071337.1 | - | 6 - 7 | 884 | 73 | ||
| BC152183.1 | - | 6 - 7 | 900 | 73 | ||
More Information
Set Membership: |
|

trnt1
Genomic Map