Assay Details
| Gene Symbol: abca4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002206.1 | NP_001002206.1 | 4 - 5 | 442 | 62 | |
| GenBank mRNA | BC074030.1 | - | 4 - 5 | 442 | 62 | |
| BC164633.1 | - | 3 - 4 | 338 | 62 | ||
More Information
Set Membership: |
|

abca4a
Genomic Map