Assay Details
| Gene Symbol: rhbdf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002228.1 | NP_001002228.1 | 16 - 17 | 2315 | 57 | |
| XM_005164335.4 | XP_005164392.1 | 17 - 18 | 2623 | 57 | ||
| GenBank mRNA | BC074097.1 | - | 16 - 17 | 2315 | 57 | |
More Information
Set Membership: |
|

rhbdf1a
Genomic Map