Assay Details
| Gene Symbol: rbp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002307.1 | NP_001002307.1 | 2 - 3 | 286 | 111 | |
| GenBank mRNA | AY619686.1 | - | 2 - 3 | 286 | 111 | |
| BC154186.1 | - | 2 - 3 | 302 | 111 | ||
More Information
Set Membership: |
|

rbp2b
Genomic Map