Assay Details
| Gene Symbol: rbp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002307.1 | NP_001002307.1 | 3 - 4 | 391 | 70 | |
| GenBank mRNA | AY619686.1 | - | 3 - 4 | 391 | 70 | |
| BC095175.1 | - | 3 - 4 | 390 | 70 | ||
| BC154186.1 | - | 3 - 4 | 407 | 70 | ||
| BC164939.1 | - | 3 - 4 | 389 | 70 | ||
More Information
Set Membership: |
|

rbp2b
Genomic Map