Assay Details
| Gene Symbol: gbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002343.1 | NP_001002343.1 | 3 - 4 | 492 | 72 | |
| XM_005171375.4 | XP_005171432.1 | 3 - 4 | 666 | 72 | ||
| XM_021479745.1 | XP_021335420.1 | 3 - 4 | 662 | 72 | ||
| GenBank mRNA | BC075920.1 | - | 3 - 4 | 492 | 72 | |
More Information
Set Membership: |
|

gbp1
Genomic Map