Assay Details
| Gene Symbol: tsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002370.1 | NP_001002370.1 | 3 - 4 | 723 | 59 | |
| GenBank mRNA | BC075886.1 | - | 3 - 4 | 723 | 59 | |
More Information
Set Membership: |
|

tsr3
Genomic Map