Assay Details
| Gene Symbol: entpd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002379.2 | NP_001002379.2 | 2 - 3 | 236 | 61 | |
| GenBank mRNA | BC075878.1 | - | 2 - 3 | 221 | 61 | |
| DT068903.1 | - | 2 - 3 | 221 | 61 | ||
More Information
Set Membership: |
|

entpd8
Genomic Map