Assay Details
| Gene Symbol: pqbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002435.1 | NP_001002435.1 | 5 - 6 | 604 | 65 | |
| GenBank mRNA | BC076131.1 | - | 5 - 6 | 604 | 65 | |
More Information
Set Membership: |
|

pqbp1
Genomic Map