Assay Details
| Gene Symbol: psmd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002436.2 | NP_001002436.2 | 4 - 5 | 711 | 71 | |
| XM_005169066.4 | XP_005169123.1 | 4 - 5 | 697 | 71 | ||
| GenBank mRNA | AI437369.1 | - | 2 - 3 | 242 | 71 | |
| BC076129.1 | - | 4 - 5 | 677 | 71 | ||
More Information
Set Membership: |
|

psmd9
Genomic Map